Learn More
Ethyl 3-ethoxyacrylate, 98%, Thermo Scientific Chemicals
£57.80 - £194.00
Chemical Identifiers
CAS | 1001-26-9 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00009863 |
InChI Key | ITQFPVUDTFABDH-AATRIKPKSA-N |
Synonym | ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate |
PubChem CID | 5366269 |
IUPAC Name | ethyl (E)-3-ethoxyprop-2-enoate |
SMILES | CCOC=CC(=O)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455428
|
Thermo Scientific Alfa Aesar
H59187.09 |
10 g |
£57.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465428
|
Thermo Scientific Alfa Aesar
H59187.18 |
50 g |
£194.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
1001-26-9 | |
144.17 | |
ITQFPVUDTFABDH-AATRIKPKSA-N | |
5366269 | |
CCOC=CC(=O)OCC |
C7H12O3 | |
MFCD00009863 | |
ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate | |
ethyl (E)-3-ethoxyprop-2-enoate |
Specifications
1001-26-9 | |
0.998 g/mL | |
76°C (169°F) | |
CH3CH2OCH=CHCOOCH2CH3 | |
MFCD00009863 | |
1721612 | |
Slightly soluble in water. | |
CCOC=CC(=O)OCC | |
144.17 | |
144.17 | |
Liquid |
Colorless | |
195°C to 196°C | |
C7H12O3 | |
1.4465 | |
UN3334 | |
ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate | |
ITQFPVUDTFABDH-AATRIKPKSA-N | |
ethyl (E)-3-ethoxyprop-2-enoate | |
5366269 | |
98% | |
Ethyl 3-ethoxyacrylate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only