missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-ethoxypropionate, 99%, Honeywell™
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Melting Point | 49.99°C |
Color | Colorless |
Density | 0.95g/mL at 25°C |
Boiling Point | 166°C |
Flash Point | 58°C (136.4°F) |
Packaging | Glass Bottle |
Linear Formula | C2H5OCH2CH2CO2C2H5 |
Refractive Index | n20/D 1.405 |
Quantity | 4 L |
UN Number | UN3272 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'