missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-formylbenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11429690
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 3-formylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-formylbenzoate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33745-47-0
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD08064275
InChI Key XZYGRRSTJVFZLS-UHFFFAOYSA-N
Synonym 3-formylbenzoic acid ethyl ester, 3-formyl-benzoic acid ethyl ester, 3-carbethoxybenzaldehyde, ethyl 3-methanoylbenzoate, acmc-20an34, 3-ethoxycarbonylbenzaldehyde
PubChem CID 11084437
IUPAC Name ethyl 3-formylbenzoate
SMILES CCOC(=O)C1=CC=CC(=C1)C=O

Specifications

Quantity 1 g
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material Ethyl 3-formylbenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.