missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-iodobenzoate, 99%, Thermo Scientific Chemicals

Product Code. 11452297
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 3-iodobenzoate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-iodobenzoate is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store in the dark. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58313-23-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00044769
InChI Key POGCXCWRMMXDAQ-UHFFFAOYSA-N
Synonym ethyl-3-iodobenzoate, 3-iodobenzoic acid ethyl ester, benzoic acid, 3-iodo-, ethyl ester, ethyl m-iodobenzoate, m-iodobenzoic acid ethyl ester, pubchem3883, ethyl-3-iodo-benzoate, acmc-1aupw, ethyl 3-iodobenzoate, rarechem al bi 0709
PubChem CID 143542
IUPAC Name ethyl 3-iodobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)I

Specifications

Density 1.66
Boiling Point 272°C
Flash Point >110°C (230°F)
Refractive Index 1.5815
Quantity 50 g
Beilstein 2089064
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 276.08
Percent Purity 99%
Chemical Name or Material Ethyl 3-iodobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'