missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-2-oxobutyrate, 95%, Thermo Scientific Chemicals

Product Code. 10381124
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20201-24-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009122
InChI Key CKTYYUQUWFEUCO-UHFFFAOYSA-N
Synonym ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate
PubChem CID 88406
IUPAC Name ethyl 3-methyl-2-oxobutanoate
SMILES CCOC(=O)C(=O)C(C)C

Specifications

Color Yellow
Density 0.9800g/mL
Boiling Point 62°C (11.0 mmHg)
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3)2CHCOCO2C2H5
Refractive Index 1.4090 to 1.413
Quantity 25 g
Beilstein 03, 683
Specific Gravity 0.98
Formula Weight 144.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 3-methyl-2-oxobutyrate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-methyl-2-oxobutyrate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'