missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-4-nitrobenzoate, 98%, Thermo Scientific™

Product Code. 11438567
Click to view available options
Quantity:
10 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30650-90-9
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00115804
InChI Key OAXJZQMFWBIRKF-UHFFFAOYSA-N
PubChem CID 2796918
IUPAC Name ethyl 3-methyl-4-nitrobenzoate
SMILES CCOC(=O)C1=CC=C(C(C)=C1)[N+]([O-])=O

Specifications

Melting Point 53°C to 55°C
Quantity 250 g
Synonym benzoic acid, 3-methyl-4-nitro-, ethyl ester, 3-methyl-4-nitrobenzoic acid ethyl ester, acmc-209zoi, ethyl 3-methyl-4-nitro-benzoate, benzoic acid,3-methyl-4-nitro-, ethyl ester
Formula Weight 209.2
Percent Purity 98%
Chemical Name or Material Ethyl 3-methyl-4-nitrobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'