missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals
£55.50 - £150.00
Chemical Identifiers
CAS | 618-98-4 |
---|---|
Molecular Formula | C9H9NO4 |
Molecular Weight (g/mol) | 195.174 |
MDL Number | MFCD00014702 |
InChI Key | MKBIJCPQTPFQKQ-UHFFFAOYSA-N |
Synonym | ethyl m-nitrobenzoate, benzoic acid, 3-nitro-, ethyl ester, ethyl-m-nitrobenzoate, m-nitrobenzoic acid, ethyl ester, benzoic acid, m-nitro-, ethyl ester, acmc-20akzc, 3-nitrobenzoic acid ethyl, benzoic acid, 3-nitro-,ethyl ester, benzoic acid, m-nitro-, ethyl ester 8ci, r---2-2-diphenylphosphino phenyl-4-phenyl-2-oxazoline |
PubChem CID | 69261 |
IUPAC Name | ethyl 3-nitrobenzoate |
SMILES | CCOC(=O)C1=CC(=CC=C1)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483334
|
Thermo Scientific Alfa Aesar
A10626.14 |
25 g |
£55.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493334
|
Thermo Scientific Alfa Aesar
A10626.22 |
100 g |
£150.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
618-98-4 | |
195.174 | |
MKBIJCPQTPFQKQ-UHFFFAOYSA-N | |
69261 | |
CCOC(=O)C1=CC(=CC=C1)[N+](=O)[O-] |
C9H9NO4 | |
MFCD00014702 | |
ethyl m-nitrobenzoate, benzoic acid, 3-nitro-, ethyl ester, ethyl-m-nitrobenzoate, m-nitrobenzoic acid, ethyl ester, benzoic acid, m-nitro-, ethyl ester, acmc-20akzc, 3-nitrobenzoic acid ethyl, benzoic acid, 3-nitro-,ethyl ester, benzoic acid, m-nitro-, ethyl ester 8ci, r---2-2-diphenylphosphino phenyl-4-phenyl-2-oxazoline | |
ethyl 3-nitrobenzoate |
Specifications
618-98-4 | |
297°C to 298°C | |
MFCD00014702 | |
14,3832 | |
MKBIJCPQTPFQKQ-UHFFFAOYSA-N | |
ethyl 3-nitrobenzoate | |
69261 | |
≥98% |
40°C to 43°C | |
C9H9NO4 | |
1912880 | |
ethyl m-nitrobenzoate, benzoic acid, 3-nitro-, ethyl ester, ethyl-m-nitrobenzoate, m-nitrobenzoic acid, ethyl ester, benzoic acid, m-nitro-, ethyl ester, acmc-20akzc, 3-nitrobenzoic acid ethyl, benzoic acid, 3-nitro-,ethyl ester, benzoic acid, m-nitro-, ethyl ester 8ci, r---2-2-diphenylphosphino phenyl-4-phenyl-2-oxazoline | |
CCOC(=O)C1=CC(=CC=C1)[N+](=O)[O-] | |
195.174 | |
195.18 | |
Ethyl 3-nitrobenzoate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-574-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only