missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-oxo-3-(4-pyridyl)propionate, 98%, Thermo Scientific Chemicals

Product Code. 15488208
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26377-17-3
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094021
InChI Key PCJNYGPKMQQCPX-UHFFFAOYSA-N
Synonym ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester
PubChem CID 2735202
IUPAC Name ethyl 3-oxo-3-pyridin-4-ylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=NC=C1

Specifications

Melting Point 53°C to 59°C
Color Brown
Density 1.155 g/mL
Refractive Index 1.51
Quantity 25 g
Formula Weight 193.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 3-oxo-3-(4-pyridyl)propionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'