missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific™

£23.11 - £279.00

Chemical Identifiers

CAS 87121-89-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD09834114
InChI Key BXBRFSMPBOTZHJ-UHFFFAOYSA-N
PubChem CID 22467070
IUPAC Name ethyl 3-oxocyclobutane-1-carboxylate
SMILES CCOC(=O)C1CC(=O)C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15498918
View Documents
Thermo Scientific Alfa Aesar
H62420.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15478918
View Documents
Thermo Scientific Alfa Aesar
H62420.03
1 g
£77.89
1g
Please to purchase this item. Need a web account? Register with us today!
15488918
View Documents
Thermo Scientific Alfa Aesar
H62420.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 3-oxocyclobutanecarboxylate is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

87121-89-9
142.154
BXBRFSMPBOTZHJ-UHFFFAOYSA-N
ethyl 3-oxocyclobutane-1-carboxylate
C7H10O3
MFCD09834114
22467070
CCOC(=O)C1CC(=O)C1

Specifications

87121-89-9
MFCD09834114
BXBRFSMPBOTZHJ-UHFFFAOYSA-N
ethyl 3-oxocyclobutane-1-carboxylate
22467070
97%
C7H10O3
ethyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid ethyl ester, 3-oxocyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 3-oxo-, ethyl ester, 3-ethoxycarbonylcyclobutanone, ethoxycarbonyl-3-cyclobutanone, ethyl3-oxocyclobutanecarboxylate, 3-ethoxycarbonyl-1-cyclobutanone, ethyl 3-oxo-cyclobutanecarboxylate
CCOC(=O)C1CC(=O)C1
142.154
142.15
Ethyl 3-oxocyclobutanecarboxylate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications