missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate, 97%, Thermo Scientific Chemicals

Product Code. 11374356
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91600-33-8
Molecular Formula C7H11F3O3
Molecular Weight (g/mol) 200.157
MDL Number MFCD00190634
InChI Key MFRIOKNLYRUYHP-UHFFFAOYSA-N
Synonym ethyl 2-methyl-3-hydroxy-4,4,4-trifluorobutyrate, ethyl 3-hydroxy-2-methyl-4,4,4-trifluorobutyrate, ethyl 2-methyl-4,4,4-trifluoro-3-hydroxybutanoate, ethyl-4,4,4-trifluor-3-hydroxy-2-methylbutanoat, ethyl 2-methyl-4,4,4-trifluoro-3-hydroxybutyrate, ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate, ethyl 3-hydroxy-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate, 4,4,4-trifluoro-3-hydroxy-2-methylbutyric acid ethyl ester
PubChem CID 533787
IUPAC Name ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutanoate
SMILES CCOC(=O)C(C)C(C(F)(F)F)O

Specifications

Boiling Point 180°C to 184°C
Quantity 1 g
Beilstein 6194166
Formula Weight 200.16
Percent Purity 97%
Chemical Name or Material Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'