missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Ethyl 4-(5-chlorocarbonyl-2-furyl)benzoate
£34.96 - £123.00
Chemical Identifiers
CAS | 333435-04-4 |
---|---|
Molecular Formula | C14H11ClO4 |
Molecular Weight (g/mol) | 278.688 |
MDL Number | MFCD02258008 |
InChI Key | OPOPFTSILPZGEL-UHFFFAOYSA-N |
Synonym | ethyl 4-5-chlorocarbonyl-2-furyl benzoate, 4-5-chlorocarbonyl-furan-2-yl-benzoic acid ethyl ester, ethyl 4-5-chlorocarbonyl furan-2-yl benzoate, ethyl 4-5-carbonochloridoyl furan-2-yl benzoate, acmc-20amvh, ethyl 4-5-carbonochloridoylfuran-2-yl benzoate, 4-5-chlorocarbonylfuran-2-yl benzoic acid ethyl ester, benzoic acid,4-5-chlorocarbonyl-2-furanyl-, ethyl ester |
PubChem CID | 3131522 |
IUPAC Name | ethyl 4-(5-carbonochloridoylfuran-2-yl)benzoate |
SMILES | CCOC(=O)C1=CC=C(C=C1)C2=CC=C(O2)C(=O)Cl |
Description
Chemical Identifiers
333435-04-4 | |
278.688 | |
OPOPFTSILPZGEL-UHFFFAOYSA-N | |
3131522 | |
CCOC(=O)C1=CC=C(C=C1)C2=CC=C(O2)C(=O)Cl |
C14H11ClO4 | |
MFCD02258008 | |
ethyl 4-5-chlorocarbonyl-2-furyl benzoate, 4-5-chlorocarbonyl-furan-2-yl-benzoic acid ethyl ester, ethyl 4-5-chlorocarbonyl furan-2-yl benzoate, ethyl 4-5-carbonochloridoyl furan-2-yl benzoate, acmc-20amvh, ethyl 4-5-carbonochloridoylfuran-2-yl benzoate, 4-5-chlorocarbonylfuran-2-yl benzoic acid ethyl ester, benzoic acid,4-5-chlorocarbonyl-2-furanyl-, ethyl ester | |
ethyl 4-(5-carbonochloridoylfuran-2-yl)benzoate |
Specifications
333435-04-4 | |
MFCD02258008 | |
Moisture sensitive | |
OPOPFTSILPZGEL-UHFFFAOYSA-N | |
ethyl 4-(5-carbonochloridoylfuran-2-yl)benzoate | |
3131522 | |
Ethyl 4-(5-chlorocarbonyl-2-furyl)benzoate |
C14H11ClO4 | |
UN3261 | |
ethyl 4-5-chlorocarbonyl-2-furyl benzoate, 4-5-chlorocarbonyl-furan-2-yl-benzoic acid ethyl ester, ethyl 4-5-chlorocarbonyl furan-2-yl benzoate, ethyl 4-5-carbonochloridoyl furan-2-yl benzoate, acmc-20amvh, ethyl 4-5-carbonochloridoylfuran-2-yl benzoate, 4-5-chlorocarbonylfuran-2-yl benzoic acid ethyl ester, benzoic acid,4-5-chlorocarbonyl-2-furanyl-, ethyl ester | |
CCOC(=O)C1=CC=C(C=C1)C2=CC=C(O2)C(=O)Cl | |
278.688 | |
278.69 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen