missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-dihydroxy-5-nitronicotinate, 97%, Thermo Scientific™

Product Code. 15437067
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6317-97-1
Molecular Formula C8H8N2O6
Molecular Weight (g/mol) 228.16
MDL Number MFCD01765414
InChI Key JBTWWCUXSHUDEJ-UHFFFAOYSA-N
Synonym ethyl 4,6-dihydroxy-5-nitronicotinate, 4,6-dihydroxy-5-nitropyridine-3-carboxylic acid ethyl ester, ethyl 4,6-dihydroxy-5-nitropyridine-3-carboxylate, 4,6-dihydroxy-5-nitronicotinic acid ethyl ester, ethyl 6-hydroxy-5-nitro-4-oxo-1h-pyridine-3-carboxylate, ethyldihydroxynitronicotinate, ethyl4,6-dihydroxy-5-nitronicotinate, ethyl 4-hydroxy-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate, 4-hydroxy-5-nitro-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylicacid, 1,6-dihydro-4-hydroxy-5-nitro-6-oxo-, ethyl ester
PubChem CID 54694073
SMILES CCOC(=O)C1=CNC(O)=C(C1=O)[N+]([O-])=O

Specifications

Melting Point 243°C to 246°C
Quantity 250 mg
IUPAC Name ethyl 4-hydroxy-5-nitro-6-oxo-1H-pyridine-3-carboxylate
Formula Weight 228.2
Percent Purity 97%
Chemical Name or Material Ethyl 4,6-dihydroxy-5-nitronicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'