missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-acetylbenzoate, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | 50°C to 55°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | CH3CH2OOCC6H4COCH3 |
Quantity | 5 g |
Formula Weight | 192.21 |
Percent Purity | 99% |
Physical Form | Crystalline Powder |
Show More |
Safety and Handling
Product Identifier
- Ethyl 4-acetylbenzoate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'