missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminobenzoate, 98%

Product Code. p-4510197 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10582161

Brand: Thermo Scientific Acros 150781000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 94-09-7 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-09-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007892
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 89.0°C to 92.0°C
Color White
Boiling Point 172°C (17.0 hPa)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula H2NC6H4CO2C2H5
Quantity 100 g
Beilstein 14,422
Merck Index 15,1088
Solubility Information Solubility in water: 1g/2500mL. Other solubilities: 1g/5mL alcohol,1g/2mL chloroform,1g/4mL ether,ca. 1g/40mL expressed almond oil or olive oil
Formula Weight 165.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-aminobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-aminobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.