missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminopiperidine-1-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 11382797
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58859-46-4
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.228
MDL Number MFCD00006484
InChI Key GQQQULCEHJQUJT-UHFFFAOYSA-N
Synonym ethyl 4-amino-1-piperidinecarboxylate, 4-amino-1-carbethoxypiperidine, unii-54815srp4z, 1-piperidinecarboxylic acid, 4-amino-, ethyl ester, ethyl 4-aminopiperidinecarboxylate, ethyl-4-amino-1-piperidine carboxylate, 4-amino-1-piperidinecarboxylic acid ethyl ester, ethyl-4-aminopiperidin-1-carboxylat, pubchem2064, acmc-209m7k
PubChem CID 100867
IUPAC Name ethyl 4-aminopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)N

Specifications

Density 1.004
Boiling Point 118°C to 120°C (6 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.483
Quantity 25 g
Beilstein 388597
Sensitivity Air Sensitive
Formula Weight 172.23
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminopiperidine-1-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'