missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-chloro-3-nitrobenzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Formula Weight | 229.62 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 4-chloro-3-nitrobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'