missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlorobutyrate, 97%, Thermo Scientific Chemicals

Product Code. 10225060
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-36-4
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.6
MDL Number MFCD00001004
InChI Key OPXNFHAILOHHFO-UHFFFAOYSA-N
Synonym ethyl 4-chlorobutyrate, butanoic acid, 4-chloro-, ethyl ester, ethyl gamma-chlorobutyrate, ethyl-4-chlorobutyrate, 4-chloro-butyric acid ethyl ester, butyric acid, 4-chloro-, ethyl ester, ethyl .gamma.-chlorobutyrate, 4-chlorobutyric acid ethyl ester, ethyl 4chlorobutyrate, acmc-1cqjw
PubChem CID 18474
IUPAC Name ethyl 4-chlorobutanoate
SMILES CCOC(=O)CCCCl

Specifications

Color Brown to Yellow
Density 1.0700g/mL
Boiling Point 186.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3CO2C2H5
Refractive Index 1.4296 to 1.4316
Quantity 5 g
Beilstein 02, 278
Specific Gravity 1.07
Solubility Information Solubility in water: immiscible
Formula Weight 150.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4-chlorobutyrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 4-chlorobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'