missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (4-fluorobenzoyl)acetate, 95%, Thermo Scientific Chemicals

Product Code. 15448398
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1999-00-4
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD03093631
InChI Key SJUXLKYJKQBZLM-UHFFFAOYSA-N
Synonym ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester
PubChem CID 2758844
IUPAC Name ethyl 3-(4-fluorophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 117°C to 120°C
Density 1.174
Refractive Index 1.504
Quantity 5 g
Formula Weight 210.2
Percent Purity 95%
Chemical Name or Material Ethyl (4-fluorobenzoyl)acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'