missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxyphenylacetate, 98%, Thermo Scientific Chemicals

Product Code. 11414877
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17138-28-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016491
InChI Key HYUPPKVFCGIMDB-UHFFFAOYSA-N
Synonym ethyl 4-hydroxyphenylacetate, ethyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-hydroxy-, ethyl ester, ethyl 4-hydroxyphenyl acetate, p-hydroxyphenyl acetic acid ethyl ester, ethyl p-hydroxyphenyl acetate, ethyl-4-hydroxyphenylacetate, pubchem12861, acetic acid, p-hydroxyphenyl-, ethyl ester
PubChem CID 28310
IUPAC Name ethyl 2-(4-hydroxyphenyl)acetate
SMILES CCOC(=O)CC1=CC=C(O)C=C1

Specifications

Melting Point 36°C to 38°C
Boiling Point 187°C to 189°C (20 mmHg)
Packaging Glass bottle
Quantity 50 g
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-hydroxyphenylacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.