missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methoxybenzoate, 98+%, Thermo Scientific Chemicals

Product Code. 11437876
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-30-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00015151
InChI Key FHUODBDRWMIBQP-UHFFFAOYSA-N
Synonym ethyl p-anisate, ethyl anisate, ethyl p-methoxybenzoate, p-anisic acid, ethyl ester, benzoic acid, 4-methoxy-, ethyl ester, 4-methoxybenzoic acid ethyl ester, benzoic acid, p-methoxy-, ethyl ester, ethyl p-anisoate, unii-kj95h2s7nm, p-anisic acid ethyl ester
PubChem CID 60979
IUPAC Name ethyl 4-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)OC

Specifications

Melting Point 7°C to 8°C
Density 1.111
Boiling Point 263°C
Flash Point 138°C (280°F)
Refractive Index 1.526
Quantity 250 g
Beilstein 2209700
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.