Learn More
Ethyl 4-methyl-4-pentenoate, 95%, Thermo Scientific Chemicals
£62.80 - £230.00
Chemical Identifiers
CAS | 4911-54-0 |
---|---|
Molecular Formula | C8H14O2 |
Molecular Weight (g/mol) | 142.2 |
MDL Number | MFCD00075160 |
InChI Key | RRTBSPBUHUUTHR-UHFFFAOYSA-N |
Synonym | ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd |
PubChem CID | 3416433 |
IUPAC Name | ethyl 4-methylpent-4-enoate |
SMILES | CCOC(=O)CCC(=C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10088309
|
Thermo Scientific Acros
427370010 |
1 g | Glass bottle | ||||||
10515470
|
Thermo Scientific Acros
427370050 |
5 g | Glass bottle |
£230.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4911-54-0 | |
142.2 | |
RRTBSPBUHUUTHR-UHFFFAOYSA-N | |
3416433 | |
CCOC(=O)CCC(=C)C |
C8H14O2 | |
MFCD00075160 | |
ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd | |
ethyl 4-methylpent-4-enoate |
Specifications
4911-54-0 | |
85.0°C (20.0 mmHg) | |
94% min. (GC) | |
CH2=C(CH3)CH2CH2C(O)OCH2CH3 | |
0.891 | |
RRTBSPBUHUUTHR-UHFFFAOYSA-N | |
ethyl 4-methylpent-4-enoate | |
3416433 | |
95% |
0.8910g/mL | |
47°C | |
C8H14O2 | |
MFCD00075160 | |
ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd | |
CCOC(=O)CCC(=C)C | |
142.2 | |
142.2 | |
Ethyl 4-methyl-4-pentenoate |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
- Ethyl 4-methyl-4-pentenoate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only