missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 98+%, Thermo Scientific Chemicals Product Code.: 11389979

Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 98+%, Thermo Scientific Chemicals

Product Code. 11389979
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11389979

Brand: Thermo Scientific Alfa Aesar H25762.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6440-02-4
Molecular Formula C5H7N3O2S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00778317
InChI Key ZUGOHJWUTNKRMG-UHFFFAOYSA-N
Synonym ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 5-amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, 1,2,3-thiadiazole-4-carboxylic acid, 5-amino-, ethyl ester, 5-amino-1,2,3 thiadiazole-4-carboxylic acid ethyl ester, acmc-20am6n, tos-bb-0200, zugohjwutnkrmg-uhfffaoysa, 5-amino-4-carbethoxy-1,2,3-thiadiazole, 5-amino-4-ethoxycarbonyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-4-carboxylicacid,5-amino-,ethylester
PubChem CID 1831000
IUPAC Name ethyl 5-aminothiadiazole-4-carboxylate
SMILES CCOC(=O)C1=C(SN=N1)N

Specifications

Melting Point 123°C to 125°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 173.19
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.