missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-amino-5-methoxyindole-2-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. 11417236
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107575-60-0
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.255
MDL Number MFCD08064224
InChI Key DIEUGINJYISDCB-UHFFFAOYSA-N
PubChem CID 10966428
IUPAC Name ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(=C(C=C2N1)N)OC

Specifications

Melting Point 167°C to 169°C
Quantity 5 g
Synonym ethyl 6-amino-5-methoxyindole-2-carboxylate, acmc-209zno, ethyl6-amino-5-methoxy-1h-indole-2-carboxylate, ethyl 6-amino-5-methoxy-1h-indole-2-carboxylate, 1h-indole-2-carboxylicacid, 6-amino-5-methoxy-, ethyl ester
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 234.25
Percent Purity 95%
Chemical Name or Material Ethyl 6-amino-5-methoxyindole-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'