missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 6-aminohexanoate, 98%, Thermo Scientific Chemicals Product Code.: 11479440

Ethyl 6-aminohexanoate, 98%, Thermo Scientific Chemicals

Product Code. 11479440
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11479440

Brand: Thermo Scientific Alfa Aesar H50296.MD

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl 6-aminohexanoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-aminohexanoate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-34-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00233495
InChI Key NJNQDCIAOXIFTB-UHFFFAOYSA-N
Synonym 6-aminohexanoic acid ethyl ester, ethyl 6-aminocaproate, ethyl-6-aminohexanoate, acmc-20aky1, ethyl-epsilon-aminocaproate, 6-aminocaproic acid ethyl ester, 6-amino-hexanoic acid ethyl ester, hexanoic acid, 6-amino-, ethyl ester
PubChem CID 406972
IUPAC Name ethyl 6-aminohexanoate
SMILES CCOC(=O)CCCCCN

Specifications

Linear Formula H2N(CH2)5CO2CH2CH3
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material Ethyl 6-aminohexanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 6-aminohexanoate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.