missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 15418735
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1027511-41-6
Molecular Formula C9H8BrN3O2
Molecular Weight (g/mol) 270.086
MDL Number MFCD10574908
InChI Key PZZTXIWJXOMKEU-UHFFFAOYSA-N
Synonym ethyl 6-bromopyrazolo 1,5-a pyrimidine-3-carboxylate, 6-bromo-3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, 6-bromopyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pubchem20934, ethyl 6-bromopyrazolo 1,5-a pyrimidine-3-carboxyla, ethyl 6-bromo-pyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl 6-bromopyrazolo 1,5-alpha pyrimidine-3-carboxylate, 6-bromo-pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester
PubChem CID 45073325
IUPAC Name ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
SMILES CCOC(=O)C1=C2N=CC(=CN2N=C1)Br

Specifications

Melting Point 144°C to 146°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 270.08
Percent Purity 98%
Chemical Name or Material Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'