missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, ≥99.5%, ACS Reagent, Honeywell™ Riedel-de Haën™
ACS Reagent,≥99.5%
£22.87 - £358.06
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
MDL Number | MFCD00009171 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
Synonym | ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
IUPAC Name | ethyl acetate |
SMILES | CCOC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15603600
|
Honeywell-Riedel-de Haen
319902-500ML |
500 mL | Glass Bottle |
£27.30
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15673590
|
Honeywell-Riedel-de Haen
319902-1L |
1 L | Glass Bottle | N/A | |||||
15683590
|
Honeywell-Riedel-de Haen
319902-2.5L |
2.5 L | Glass Bottle | N/A | |||||
15623600
|
Honeywell-Riedel-de Haen
319902-6X500ML |
6 x 500 mL | Glass Bottle |
£163.61
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15638360
|
Honeywell-Riedel-de Haen
319902-4L |
4 L | Glass Bottle | N/A | |||||
15613600
|
Honeywell-Riedel-de Haen
319902-6X1L |
6 x 1 L | Glass Bottle | N/A | |||||
15693590
|
Honeywell-Riedel-de Haen
319902-20L |
20 L | Stainless Steel Can | N/A | |||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
141-78-6 | |
88.106 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
8857 | |
ethyl acetate |
C4H8O2 | |
MFCD00009171 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CHEBI:27750 | |
CCOC(=O)C |
Specifications
-84°C | |
0.902g/mL at 25°C | |
2.2 to 11.5% (38°F) | |
801°F | |
CH3COOC2H5 | |
73mmHg (20°C) | |
506104 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11g/mol | |
ACS Reagent |
141-78-6 | |
77.5°C | |
−3°C (26.6°F) | |
C4H8O2 | |
MFCD00009171 | |
UN1173 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 | |
≥99.5% | |
Ethyl Acetate |
Safety and Handling
P210-P261-P305 + P351 + P338
H225-H319-H336
missing translation for 'dotInformation' : Class 3 - PG 2 - Ethyl acetate
missing translation for 'einecsNumber' : 205-500-4
missing translation for 'rtecsNumber' : AH5425000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ethyl acetate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethyl acetate