missing translation for 'onlineSavingsMsg'
Learn More
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals Product Code.: 10052593

Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals

Product Code. 10052593
2.5 L, Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10052593

Brand: Thermo Scientific Acros 423680025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic esterShow MoreShow Less
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Density 0.9020g/mL
Boiling Point 75.0°C to 78.0°C
Quantity 2.5 L
Flash Point −4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3710 to 1.3740
Assay Percent Range 99.6%
Residue after Evaporation 0.003% max.
Specific Gravity 0.902
Titratable Acid 0.0009 meq/g max.
Linear Formula CH3CO2C2H5
Viscosity 0.45 mPa.s (20°C)
Beilstein 02, 125
Water 0.2% max. (K.F.)
Merck Index 15, 3811
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Percent Purity 99.6%
Grade ACS Reagent
Chemical Name or Material Ethyl acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2412165 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10589282,423685000,423680250,423680010,423680025
Lot Number2417257 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10589282,423685000,423680250,423680010,423680025
Lot Number2409982 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10589282,423685000,423680250,423680010,423680025
Lot Number2406494 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10589282,423685000,423680250,423680010,423680025
Lot Number2173435 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number10589282,423685000,423680250,423680010,423680025
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.