missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, 99.9%, Extra Dry, AcroSeal™

Product Code. p-216946 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10469252

Brand: Thermo Scientific Acros 326901000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Color Colorless
Density 0.9020g/mL
Boiling Point 75.0°C to 78.0°C
Quantity 100 mL
Flash Point −4°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3710 to 1.3740
Assay Percent Range 99.9%
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.902
Linear Formula CH3CO2C2H5
Viscosity 0.45 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Percent Purity 99.9%
Grade Extra Dry
Chemical Name or Material Ethyl acetate, Water <50ppm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.