missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, 99+%, extra pure

Product Code. 10376500 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
This item is not returnable. View return policy

Product Code. 10376500

Brand: Thermo Scientific Acros 149470250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Density 0.9020g/mL
Boiling Point 75.0°C to 78.0°C
Quantity 25 L
Flash Point −4°C
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.3710 to 1.3740
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.902
Linear Formula CH3CO2C2H5
Vapor Pressure Vapor Pressure: 103mbar at 20°C
Viscosity 0.45 mPa.s (20°C)
Beilstein 02, 125
Water 0.05% max. (K.F.)
Merck Index 15, 3811
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Ethyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.