missing translation for 'onlineSavingsMsg'
Learn More
Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals Product Code.: 11327458

Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals

Product Code. 11327458
4 x 1 L
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11327458

Brand: Thermo Scientific Alfa Aesar 031344.M1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.902 g/mL
Boiling Point 77°C
Quantity 4 x 1 L
Flash Point -3°C (26°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.372
Linear Formula CH3CO2CH2CH3
UN Number UN1173
Beilstein 506104
Merck Index 14,3757
Solubility Information Miscible with ethanol,acetone,diethyl ether and benzene.
Formula Weight 88.11
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Ethyl acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L005 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number031344.M1,031344.AP,031344.K7,031344.K2,031344.M4
Lot NumberN14L031 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number031344.M1,031344.AP,031344.K7,031344.K2,031344.M4
Lot NumberM31L047 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog Number031344.M1,031344.AP,031344.K7,031344.K2,031344.M4
Lot NumberY25K007 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number031344.M1,031344.AP,031344.K7,031344.K2,031344.M4
Lot NumberX04K034 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number031344.M1,031344.AP,031344.K7,031344.K2,031344.M4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.