missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Melting Point | -83°C |
CAS | 141-78-6 |
Color | Max. 10 (APHA) |
Density | 0.902 g/cm³ (20°C) |
Boiling Point | 77.1°C (1013 hPa) |
Quantity | 1 L |
Flash Point | -4°C |
Packaging | Glass Bottle |
Assay Percent Range | min. 99.5% |
Residue after Evaporation | max. 0.003% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Safety and Handling


Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethyl acetate, BAKER ANALYZED™ ACS, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction