missing translation for 'onlineSavingsMsg'
Learn More
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10429750

Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10429750
2.5 L, Plastic-coated Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
This item is not returnable. View return policy

Product Code. 10429750

Brand: Fisher Chemical E/0900/PB17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C4H8O2, CAS Number-141-78-6, acetic acid ethyl ester, acetic acid, ethyl ester, acetic ester, acetic ether, acetidin, acetoxyethane, ethylacetate, ethyl acetic ester, ethyl ethanoate, vinegar naphtha, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
MDL Number 9171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate

Specifications

Melting Point -83.5°C
Color Colorless
Boiling Point 75°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 103mbar at 20°C
Viscosity 0.45cp at 20°C
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
SMILES CCOC(=O)C
Molecular Weight (g/mol) 88.106
Formula Weight 88.11g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.