missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riedel-de Haën Ethyl Acetate CHROMASOLV™
Click to view available options
Quantity:
7 L
Description
- For HPLC and organic synthesis.
- High-purity, multipurpose solvents.
- High lot-to-lot consistency.
Specifications
pH | No data available |
Melting Point | -84°C |
CAS | 141-78-6 |
CAS Min % | ≥90.00% |
Color | Colorless |
CAS Max % | ≤100.00% |
Density | 0.901 g/cm3 |
Boiling Point | 77°C (1,013 hPa) |
Quantity | 7 L |
Flash Point | -4°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'