missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, for HPLC, Fisher Chemical™

C4H8O2, CAS Number-141-78-6, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate, -83.5 deg.C, 9171, 88.106, Amber glass bottle, Liquid, 8857, CCOC(-O)C, 3.04, 103mbar at 20 deg.C, 0.45cp at 20 deg.C

£55.80 - £117.00

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
MDL Number 9171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10724181
View Documents Promotion Details
Fisher Chemical
E/0906/15
1 L Amber glass bottle
£55.80
1 L
Please to purchase this item. Need a web account? Register with us today!
10456870
View Documents Promotion Details
Fisher Chemical
E/0906/17
2.5 L Amber glass bottle
£80.30
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10040140
View Documents Promotion Details
Fisher Chemical
E/0906/PB17
2.5 L Plastic Coated Glass bottle
£117.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C4H8O2, CAS Number-141-78-6, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate, -83.5 deg.C, 9171, 88.106, Amber glass bottle, Liquid, 8857, CCOC(-O)C, 3.04, 103mbar at 20 deg.C, 0.45cp at 20 deg.C

Specifications

Chemical Identifiers

141-78-6
9171
8857
ethyl acetate
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
CHEBI:27750

Specifications

141-78-6
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11g/mol
0.45cp at 20°C
-83.5°C
Liquid
Colorless
9171
CCOC(=O)C
88.106
CHEBI:27750
103mbar at 20°C
3.04
75°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl acetate

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications