missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, HPLC Grade, J.T. Baker™
High purity solvents for use in analytical and preparative separations
£265.00 - £436.00
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
Synonym | ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
IUPAC Name | ethyl acetate |
SMILES | CCOC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15496051
|
Avantor J.T.Baker
9282-02 |
1 L | Narrow Mouth Amber Glass Bottle |
£265.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15443224
|
Avantor J.T.Baker
9282-03 |
4 L | Narrow Mouth Amber Glass Bottle |
£436.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
Chemical Identifiers
141-78-6 | |
88.106 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CHEBI:27750 | |
CCOC(=O)C |
C4H8O2 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
8857 | |
ethyl acetate |
Specifications
141-78-6 | |
1L = 0.90kg | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 | |
HPLC |
−4.00°C (− 39.2°F) | |
C4H8O2 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11 | |
Ethyl Acetate |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethyl Acetate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .