missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl acetate, puriss. p.a., ≥99.9% (GC), Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -84°C |
CAS | 141-78-6 |
Molecular Formula | C4H8O2 |
Linear Formula | CH3COOC2H5 |
MDL Number | MFCD00009171 |
UN Number | UN1173 |
Beilstein | 506104 |
Synonym | ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
SMILES | CCOC(=O)C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'