missing translation for 'onlineSavingsMsg'
Learn More
Ethyl bromofluoroacetate, 97%, Thermo Scientific Chemicals Product Code.: 11417184

Ethyl bromofluoroacetate, 97%, Thermo Scientific Chemicals

Product Code. 11417184
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11417184

Brand: Thermo Scientific Alfa Aesar B21579.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl bromofluoroacetate is used as an intermediate useful in the synthesis of polyfluoroalkyl-substituted fluoro enol ethers and N-protected α-fluoroglycines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromofluoroacetate is used as an intermediate useful in the synthesis of polyfluoroalkyl-substituted fluoro enol ethers and N-protected α-fluoroglycines.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-55-8
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.992
MDL Number MFCD00042095
InChI Key ULNDTPIRBQGESN-UHFFFAOYSA-N
Synonym ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3tShow MoreShow Less
PubChem CID 2733407
IUPAC Name ethyl 2-bromo-2-fluoroacetate
SMILES CCOC(=O)C(F)Br

Specifications

Density 1.54
Boiling Point 154°C
Flash Point 70°C (158°F)
Refractive Index 1.4255
Quantity 25 g
UN Number UN2810
Solubility Information It is soluble in water.
Formula Weight 184.99
Percent Purity 97%
Chemical Name or Material Ethyl bromofluoroacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10206995 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog NumberB21579,B21579.14,B21579.06
Lot NumberJ13T048 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21579.14,B21579.06
Lot NumberG15T002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21579.14,B21579.06
Lot Number10206995 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21579.14,B21579.06
Lot Number10186667 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21579.14,B21579.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl bromofluoroacetate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.