missing translation for 'onlineSavingsMsg'
Learn More

Ethyl butyrate, 99%, Thermo Scientific Chemicals

Product Code. 10254670
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-54-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009394
InChI Key OBNCKNCVKJNDBV-UHFFFAOYSA-N
Synonym ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427
PubChem CID 7762
IUPAC Name ethyl butanoate
SMILES CCCC(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Melting Point -93.3°C
Color Colorless
Density 0.8700g/mL
Boiling Point 120.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CO2C2H5
Refractive Index 1.3910 to 1.3930
Quantity 5 mL
Beilstein 02, 270
Merck Index 15, 3830
Specific Gravity 0.87
Solubility Information Solubility in water: practically insoluble
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl butyrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl butyrate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.