Learn More
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals
£20.30 - £288.00
Chemical Identifiers
CAS | 105-39-5 |
---|---|
MDL Number | MFCD00000932 |
InChI Key | VEUUMBGHMNQHGO-UHFFFAOYSA-N |
Synonym | ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf |
PubChem CID | 7751 |
IUPAC Name | ethyl 2-chloroacetate |
SMILES | CCOC(=O)CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10733464
|
Thermo Scientific Acros
118222500 |
250 mL | Glass bottle |
£20.30
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783851
|
Thermo Scientific Acros
118220010 |
1 L | Glass bottle |
£42.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657262
|
Thermo Scientific Acros
118220025 |
2.5 L | Glass bottle |
£85.60
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974921
|
Thermo Scientific Acros
118220100 |
10 L | Plastic drum |
£288.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-39-5 | |
VEUUMBGHMNQHGO-UHFFFAOYSA-N | |
7751 | |
CCOC(=O)CCl |
MFCD00000932 | |
ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf | |
ethyl 2-chloroacetate |
Specifications
105-39-5 | |
Colorless | |
142.0°C to 145.0°C | |
Authentic | |
C4H7ClO2 | |
1.4200 to 1.4220 | |
02,197 | |
15,3838 | |
ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf | |
VEUUMBGHMNQHGO-UHFFFAOYSA-N | |
ethyl 2-chloroacetate | |
7751 | |
122.55 | |
Liquid |
-26.0°C | |
1.1500g/mL | |
53°C | |
98.5% min. (GC) | |
ClCH2CO2C2H5 | |
MFCD00000932 | |
13,348 | |
1.15 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone | |
CCOC(=O)CCl | |
122.55 | |
1.27 mPa.s (20°C) | |
99% | |
Ethyl chloroacetate |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-294-
- Ethyl chloroacetate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Flammable liquid Category 3
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only