missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl chloroformate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
Specifications
Specifications
Melting Point | -81.0°C |
Density | 1.1350g/mL |
Boiling Point | 94.0°C |
Flash Point | 10°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C3H5ClO2 |
Linear Formula | ClCO2C2H5 |
Refractive Index | 1.3950 to 1.3960 |
Quantity | 2.5 L |
Show More |
Safety and Handling
Product Identifier
- Ethyl chloroformate
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction