missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroformate, 97%, Thermo Scientific Chemicals

£22.21 - £261.30

Chemical Identifiers

CAS 541-41-3
InChI Key RIFGWPKJUGCATF-UHFFFAOYSA-N
Synonym ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
PubChem CID 10928
IUPAC Name ethyl carbonochloridate
SMILES CCOC(=O)Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10266190
View Documents
Thermo Scientific Acros
181080050
5 mL Glass bottle
£22.91
5mL
Please to purchase this item. Need a web account? Register with us today!
10167613
View Documents Promotion Details
Thermo Scientific Acros
181081000
100 mL Glass bottle
£22.21
100mL
Please to purchase this item. Need a web account? Register with us today!
10407950
View Documents
Thermo Scientific Acros
181085000
500 mL Glass bottle
£72.66
500mL
Please to purchase this item. Need a web account? Register with us today!
11944891
View Documents
Thermo Scientific Acros
181080025
2.5 L Glass bottle
£261.30
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

541-41-3
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
ethyl carbonochloridate
RIFGWPKJUGCATF-UHFFFAOYSA-N
10928
CCOC(=O)Cl

Specifications

541-41-3
1.1350g/mL
10°C
C3H5ClO2
1.3950 to 1.3960
01,364; 02,193; 04,228; 05,294; 07,147; 09,214; 12,223
1.135
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in benzene, chloroform and ether, soluble in most organic solvents
CCOC(=O)Cl
108.52
0.46 mPa.s (20°C)
97%
-81.0°C
94.0°C
96% min. (GC)
ClCO2C2H5
03,1
15,3839
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
RIFGWPKJUGCATF-UHFFFAOYSA-N
ethyl carbonochloridate
10928
108.52
Ethyl chloroformate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 208-778-5

Product Identifier
  • Ethyl chloroformate

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only