Learn More
Ethyl difluoroacetate, 98%, Thermo Scientific Chemicals
£55.20 - £655.00
Chemical Identifiers
CAS | 454-31-9 |
---|---|
Molecular Formula | C4H6F2O2 |
Molecular Weight (g/mol) | 124.087 |
MDL Number | MFCD00013578 |
InChI Key | GZKHDVAKKLTJPO-UHFFFAOYSA-N |
Synonym | ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat |
PubChem CID | 9961 |
IUPAC Name | ethyl 2,2-difluoroacetate |
SMILES | CCOC(=O)C(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443534
|
Thermo Scientific Alfa Aesar
A11215.06 |
5 g |
£55.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453534
|
Thermo Scientific Alfa Aesar
A11215.14 |
25 g |
£194.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463534
|
Thermo Scientific Alfa Aesar
A11215.22 |
100 g |
£655.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.
Solubility
Slightly soluble in water
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
454-31-9 | |
124.087 | |
GZKHDVAKKLTJPO-UHFFFAOYSA-N | |
9961 | |
CCOC(=O)C(F)F |
C4H6F2O2 | |
MFCD00013578 | |
ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat | |
ethyl 2,2-difluoroacetate |
Specifications
454-31-9 | |
98°C to 99°C | |
C4H6F2O2 | |
MFCD00013578 | |
1748597 | |
Slightly soluble in water | |
CCOC(=O)C(F)F | |
124.087 | |
124.09 | |
Ethyl difluoroacetate |
1.242 | |
25°C (77°F) | |
1.3475 | |
UN2924 | |
ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat | |
GZKHDVAKKLTJPO-UHFFFAOYSA-N | |
ethyl 2,2-difluoroacetate | |
9961 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 207-223-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only