missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl dodecanoate, 98%, Thermo Scientific Chemicals
£25.00 - £355.00
Chemical Identifiers
CAS | 106-33-2 |
---|---|
Molecular Formula | C14H28O2 |
Molecular Weight (g/mol) | 228.38 |
MDL Number | MFCD00015065 |
InChI Key | MMXKVMNBHPAILY-UHFFFAOYSA-N |
Synonym | ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k |
PubChem CID | 7800 |
ChEBI | CHEBI:87427 |
IUPAC Name | ethyl dodecanoate |
SMILES | CCCCCCCCCCCC(=O)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473484
|
Thermo Scientific Alfa Aesar
A11188.AE |
100 mL |
£25.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483484
|
Thermo Scientific Alfa Aesar
A11188.AP |
500 mL |
£86.70
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493484
|
Thermo Scientific Alfa Aesar
A11188.0F |
2500 mL |
£355.00
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-33-2 | |
228.38 | |
MMXKVMNBHPAILY-UHFFFAOYSA-N | |
7800 | |
ethyl dodecanoate |
C14H28O2 | |
MFCD00015065 | |
ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k | |
CHEBI:87427 | |
CCCCCCCCCCCC(=O)OCC |
Specifications
106-33-2 | |
0.862 | |
138°C (280°F) | |
C14H28O2 | |
MFCD00015065 | |
14,3818 | |
MMXKVMNBHPAILY-UHFFFAOYSA-N | |
ethyl dodecanoate | |
7800 | |
228.38 | |
Ethyl dodecanoate |
-2°C | |
268°C to 270°C | |
Fruit-like | |
1.4315 | |
1769671 | |
ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k | |
CCCCCCCCCCCC(=O)OCC | |
228.38 | |
CHEBI:87427 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 203-386-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only