Learn More
Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals
£46.33 - £168.00
Chemical Identifiers
CAS | 5205-39-0 |
---|---|
Molecular Formula | C7H11ClO3 |
Molecular Weight (g/mol) | 178.612 |
MDL Number | MFCD00013659 |
InChI Key | KKJAQUGGQMCNJY-UHFFFAOYSA-N |
Synonym | ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester |
PubChem CID | 78881 |
IUPAC Name | ethyl 5-chloro-5-oxopentanoate |
SMILES | CCOC(=O)CCCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435768
|
Thermo Scientific Alfa Aesar
H59443.AA |
5 mL |
£46.33
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445768
|
Thermo Scientific Alfa Aesar
H59443.AC |
25 mL |
£168.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
5205-39-0 | |
178.612 | |
KKJAQUGGQMCNJY-UHFFFAOYSA-N | |
78881 | |
CCOC(=O)CCCC(=O)Cl |
C7H11ClO3 | |
MFCD00013659 | |
ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester | |
ethyl 5-chloro-5-oxopentanoate |
Specifications
5205-39-0 | |
68°C to 70°C (0.07 mmHg) | |
ClCO(CH2)3COOC2H5 | |
MFCD00013659 | |
1099817 | |
Soluble in water. | |
CCOC(=O)CCCC(=O)Cl | |
178.612 | |
178.61 | |
Liquid |
1.139 | |
C7H11ClO3 | |
1.443 | |
UN3265 | |
ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester | |
KKJAQUGGQMCNJY-UHFFFAOYSA-N | |
ethyl 5-chloro-5-oxopentanoate | |
78881 | |
97% | |
Ethyl glutaryl chloride |
Safety and Handling
GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 226-000-2
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only