missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl glyoxalate, technical, ∼50% in toluene, Thermo Scientific Chemicals
£120.00 - £198.00
Chemical Identifiers
CAS | 924-44-7 |
---|---|
Molecular Formula | C4H6O3 |
Molecular Weight (g/mol) | 102.09 |
InChI Key | DBPFRRFGLYGEJI-UHFFFAOYSA-N |
Synonym | ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester |
PubChem CID | 70211 |
ChEBI | CHEBI:53275 |
IUPAC Name | ethyl 2-oxoacetate |
SMILES | CCOC(=O)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10766265
|
Thermo Scientific Acros
425765000 |
500 g |
£120.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10103384
|
Thermo Scientific Acros
425760010 |
1 kg |
£198.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
924-44-7 | |
102.09 | |
ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester | |
CHEBI:53275 | |
CCOC(=O)C=O |
C4H6O3 | |
DBPFRRFGLYGEJI-UHFFFAOYSA-N | |
70211 | |
ethyl 2-oxoacetate |
Specifications
924-44-7 | |
110°C | |
Glass Bottle | |
HCOCO2CH2CH3 | |
ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester | |
CCOC(=O)C=O | |
102.09 | |
CHEBI:53275 | |
Technical |
1.0320g/mL | |
7°C | |
C4H6O3 | |
1.032 | |
DBPFRRFGLYGEJI-UHFFFAOYSA-N | |
ethyl 2-oxoacetate | |
70211 | |
102.09 | |
Ethyl glyoxalate, Approx. 50% in toluene |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-105-3
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethyl glyoxalate, ca 50% in toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only