missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hippurate, 96%, Thermo Scientific Chemicals

Product Code. 11476486
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-53-2
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00026890
InChI Key PTXRQIPIELXJFH-UHFFFAOYSA-N
Synonym ethyl hippurate, ethyl n-benzoylglycinate, n-benzoylglycine ethyl ester, ethyl 2-phenylformamido acetate, benzoic amide, n-ethoxycarbonyl methyl, ethylhippurate, ethyl benzoylaminoacetate, acmc-1buir, hippuric acid ethyl ester, maybridge1_006087
PubChem CID 226558
IUPAC Name ethyl 2-benzamidoacetate
SMILES CCOC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Quantity 100 g
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material Ethyl hippurate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'