Learn More
Ethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
£36.50 - £71.40
Chemical Identifiers
CAS | 97-63-2 |
---|---|
Molecular Formula | C6H10O2 |
Molecular Weight (g/mol) | 114.14 |
MDL Number | MFCD00009161 |
InChI Key | SUPCQIBBMFXVTL-UHFFFAOYSA-N |
Synonym | ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester |
PubChem CID | 7343 |
IUPAC Name | ethyl 2-methylprop-2-enoate |
SMILES | CCOC(=O)C(=C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15720839
|
Thermo Scientific Acros
215835000 |
500 g |
£36.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10092920
|
Thermo Scientific Acros
215830010 |
1 kg |
£71.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-63-2 | |
114.14 | |
SUPCQIBBMFXVTL-UHFFFAOYSA-N | |
7343 | |
CCOC(=O)C(=C)C |
C6H10O2 | |
MFCD00009161 | |
ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester | |
ethyl 2-methylprop-2-enoate |
Specifications
97-63-2 | |
Colorless | |
118.0°C to 119.0°C | |
Authentic | |
Glass bottle | |
H2C=C(CH3)CO2C2H5 | |
MFCD00009161 | |
0.91 | |
Solubility in water: 4g/L in water (20°C). Other solubilities: soluble in acetone,slightly soluble in chloroform,miscible in ether and ethanol | |
CCOC(=O)C(=C)C | |
10 to 20ppm MEHQ | |
7343 | |
114.14 | |
Liquid |
-75.0°C | |
0.9100g/mL | |
15°C | |
98.5% min. (GC) | |
C6H10O2 | |
1.4130 to 1.4150 | |
02, 423 | |
ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester | |
SUPCQIBBMFXVTL-UHFFFAOYSA-N | |
ethyl 2-methylprop-2-enoate | |
114.14 | |
0.63 mPa.s (20°C) | |
99% | |
Ethyl methacrylate, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-597-5
missing translation for 'rtecsNumber' : OZ4550000
missing translation for 'tsca' : TSCA
RUO – Research Use Only