Learn More
Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals
£25.70 - £474.00
Chemical Identifiers
CAS | 62-50-0 |
---|---|
Molecular Formula | C3H8O3S |
Molecular Weight (g/mol) | 124.154 |
MDL Number | MFCD00007559 |
InChI Key | PLUBXMRUUVWRLT-UHFFFAOYSA-N |
Synonym | ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid |
PubChem CID | 6113 |
ChEBI | CHEBI:23994 |
IUPAC Name | ethyl methanesulfonate |
SMILES | CCOS(=O)(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11478930
|
Thermo Scientific Alfa Aesar
A12938.06 |
5 g |
£25.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499763
|
Thermo Scientific Alfa Aesar
A12938.09 |
10 g |
£46.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409773
|
Thermo Scientific Alfa Aesar
A12938.18 |
50 g |
£168.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419773
|
Thermo Scientific Alfa Aesar
A12938.30 |
250 g |
£474.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.
Solubility
Miscible with water.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS available online at www.alfa.com.Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
62-50-0 | |
124.154 | |
PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
6113 | |
ethyl methanesulfonate |
C3H8O3S | |
MFCD00007559 | |
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
CHEBI:23994 | |
CCOS(=O)(=O)C |
Specifications
62-50-0 | |
85°C to 86°C (10 mmHg) | |
99% | |
C3H8O3S | |
MFCD00007559 | |
773969 | |
14,3827 | |
Miscible with water. | |
CCOS(=O)(=O)C | |
124.154 | |
CHEBI:23994 | |
99% |
1.17 | |
100°C (212°F) | |
Fruit-like | |
1.418 | |
UN2810 | |
Moisture sensitive | |
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
ethyl methanesulfonate | |
6113 | |
124.16 | |
Ethyl methanesulfonate |
Safety and Handling
GHS H Statement
H340-H351-H361-H302
May cause genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H340-H351
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 200-536-7
missing translation for 'rtecsNumber' : PB2100000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Moisture Sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
Precautionary Statement
- P201-Obtain special instructions before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only