missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
39.34 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Description
C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -87°C |
Color | Colorless |
Boiling Point | 80°C |
Packaging | Amber glass bottle |
Quantity | 500 mL |
Formula Weight | 72.11g/mol |
Physical Form | Liquid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 2-Butanone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P240-Ground/bond container and receiving equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'