missing translation for 'onlineSavingsMsg'
Learn More

Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate, 98+%, Thermo Scientific Chemicals

Product Code. 15432126
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38202-27-6
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.358
MDL Number MFCD00009244
InChI Key KQCBSWBQAXTILK-WYMLVPIESA-N
Synonym ethyl o-mesitylsulfonylacetohydroxamate, ethyl n-mesitylsulfonyl oxyacetimidate, ethyl o-methylsulphonylacetohydroxamate, ethyl o-2-mesitylenesulfonyl acethydroxamate, o-mesitylsulfonylacetohydroxamic acid ethyl ester, ethyl 1z-n-mesitylsulfonyl oxy ethanimidoate, ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate, kqcbswbqaxtilk-owbhpgmisa-n, z-ethyl n-mesitylsulfonyloxyacetimidate, z-ethyl n-mesitylsulfonyl oxyacetimidate
PubChem CID 6282444
IUPAC Name ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate
SMILES CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C

Specifications

Melting Point 54°C to 56°C
Quantity 1 g
Beilstein 2144156
Formula Weight 285.36
Percent Purity ≥98%
Chemical Name or Material Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.